missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Xylene Substitute Mountant

Employ all routine xylene-substitute clearing agents with the Epredia™ Xylene Substitute Mountant.

$110.50 - $701.75

Specifications

Product Type Xylene Substitute
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
2860057
View Documents Promotion Details
Epredia™
1900231
60 mL
Each for $110.50
Only null left
 
1900233
View Documents Promotion Details
Epredia™
1900233
120 mL
Each for $118.64
Only null left
 
9999122
View Documents Promotion Details
Epredia™
9999122
500 mL
Case of 2 Each for $701.75
Only null left
 
Description

Description

  • Permanent, organic mounting medium
  • Toluene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (120-265 cps at 24°C)
Specifications

Specifications

Xylene Substitute
Videos
SDS
Documents

Documents