missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Xylene Substitute Mountant
Employ all routine xylene-substitute clearing agents with the Epredia™ Xylene Substitute Mountant.
Supplier: Epredia™ 1900231
Description
- Permanent, organic mounting medium
- Toluene base
- Refractive index: 1.495 ±0.005 (dry)
- Viscosity: Medium (120-265 cps at 24°C)
Chemical Identifiers
| Xylene Substitute |
Specifications
| Xylene Substitute | |
| 60 mL |