missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH3RF1 Monoclonal Antibody (K1E030_5E8), Invitrogen™

Catalog No. MA555676
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555676 50 μL
MA556056 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555676 Supplier Invitrogen™ Supplier No. MA555676
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GNSSATKPDK DSKKEKKGLL KLLSGASTKR KPRVSPPASP TLEVELGSAE LPLQGAVGPE LPPGGGHGRA GSCPVDGDGP VTTAVAGAAL AQDAFHRKAS SLDSAVPIAP PPRQACSSLG PVLNESRPVV CERHRVVVSY PPQSEAELEL KEGDIVFVHK KREDGWFKGT LQRNGKTGLF

This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via the RING-finger, the encoded protein has been shown to function as an ubiquitin-protein ligase involved in protein sorting at the trans-Golgi network. The encoded protein may also act as a scaffold for the c-Jun N-terminal kinase signaling pathway, facilitating the formation of a functional signaling module.

Specifications

Antigen SH3RF1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K1E030_5E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SH3RF1
Gene Accession No. Q69ZI1, Q7Z6J0
Gene Alias 2200003J05Rik; E3 ubiquitin-protein ligase SH3RF1; FLJ21602; KIAA1494; plenty of SH3 domains; plenty of SH3s; Posh; Posh1; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative E3 ubiquitin-protein ligase SH3RF1; E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; R75531; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; RNF142; SH3 domain containing ring finger 1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2; SH3MD2; Sh3rf1
Gene Symbols SH3RF1
Host Species Mouse
Immunogen Recombinant human SH3RF1 (immunogen range is 701-880).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57630, 59009
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less