missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH2D2A Monoclonal Antibody (OTI4F3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525894
Description
SH2D2A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.Specifications
| SH2D2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NP31 | |
| Sh2d2a | |
| Full length human recombinant protein of SH2D2A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4F3 | |
| Unconjugated | |
| Sh2d2a | |
| F2771; Lad; Lck-associated adapter protein; p56Lck-associated adapter protein; Ribp; Rlk/Itk-binding protein; SCAP; SH2 domain containing 2A; SH2 domain protein 2A; SH2 domain-containing adapter protein; SH2 domain-containing protein 2A; SH22A; SH2D2A; T cell specific adapter protein TSAd; T cell-specific adapter protein; T lymphocyte specific adaptor protein; Tsad; VEGF receptor-associated protein; VRAP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9047 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |