missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SGK (SGK1), GST Tag Recombinant Protein

Catalog No. PR7358A
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7358A 100 μg
PV3818 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR7358A Supplier Invitrogen™ Supplier No. PR7358A
Only null left

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

SGK (SGK1) is a serine/threonine kinase that may be a transcriptional target for p38/MAPK in hepatoma cells. SGK plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.

Specifications

Accession Number O00141
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 0.5mM EDTA, 150mM NaCl, 2mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6446
Molecular Weight (g/mol) 69.7 kDa
Name Human SGK (SGK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3
Common Name SGK1
Gene Symbol SGK1
KinaseFamily SGK Kinase Family
Protein Length 60-431
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 60-431
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: AGC Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less