missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150083
Description
SFTPA2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication.Specifications
| SFTPA2 | |
| Polyclonal | |
| Unconjugated | |
| SFTPA2 | |
| 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC5; collectin 5; Collectin-5; PSAP; PSPA; PSP-A; Pulmonary surfactant-associated protein A2; SFTP1; SFTPA; SFTPA2; SFTPA2B; SP-2A; SP-2A beta; SP-2A gamma; SP-A; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant protein A2; surfactant, pulmonary-associated protein A2A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 729238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8IWL1 | |
| SFTPA2 | |
| Recombinant Surfactant Associated Protein A2 (SPA2) (Glu21-Phe114). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |