missing translation for 'onlineSavingsMsg'
Learn More

SFRS18/SRrp130 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560823
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560823 Supplier Bethyl Laboratories Inc Supplier No. A301609A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

SFRS18 is a protein coding gene.

Specifications

Antigen SFRS18/SRrp130
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PNISR
Gene Accession No. Q8TF01
Gene Alias C6orf111, HSPC306, RP11-98I9.2, SFRS18, SRrp130, bA98I9.2, SR rich protein, SR-related protein, SR-rich protein, arginine/serine-rich protein PNISR, serine-arginine-rich-splicing regulatory protein 130, serine/arginine-rich-splicing regulatory protein 130, splicing factor, arginine/serine-rich 130, splicing factor, argi
Gene Symbols PNISR
Host Species Rabbit
Immunogen Between 200 and 250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25957
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats