missing translation for 'onlineSavingsMsg'
Learn More

SFRS17A Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564566
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501564566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501564566 Supplier Bethyl Laboratories Inc Supplier No. A303218A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.

Specifications

Antigen SFRS17A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AKAP17A
Gene Accession No. Q02040
Gene Alias RP13-297E16.1, 721P, AKAP-17A, CCDC133, CXYorf3, DXYS155E, PRKA17A, SFRS17A, XE7, XE7Y, A-kinase anchor protein 17A, B-lymphocyte surface antigen, protein kinase A-anchoring protein 17A, pseudoautosomal gene XE7, splicing factor, arginine/serine-rich 17A
Gene Symbols AKAP17A
Host Species Rabbit
Immunogen Between 575 and 625
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8227
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less