missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SF3A3 Recombinant Rabbit Monoclonal Antibody (23GB4775)

Catalog No. MA552741
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552741 20 μL
MA552740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552741 Supplier Invitrogen™ Supplier No. MA552741
Only null left

Rabbit Recombinant Monoclonal Antibody

SF3A3 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20.

Specifications

Antigen SF3A3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4775
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SF3A3
Gene Accession No. Q12874, Q9D554
Gene Alias 4930512K19Rik; 60kDa; hypothetical protein LOC523250; LRRGT00093; pre-mRNA splicing factor SF3a (60kD); PRP9; PRPF9; SAP 61; Sap61; SF3A3; SF3a60; spliceosome associated protein 61; spliceosome-associated protein 61; splicesome-associated factor 61; splicing factor 3A subunit 3; splicing factor 3a subunit 3-like protein; splicing factor 3a, subunit 3; splicing factor 3a, subunit 3, 60kD; splicing factor 3a, subunit 3, 60kDa; wu:fi15a06; wu:fj94f01; zgc:110227
Gene Symbols SF3A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SF3A3 (1-28AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10946, 313583, 75062
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less