missing translation for 'onlineSavingsMsg'
Learn More

SF1 Antibody, ProSci

Catalog No. 16832140
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16832140 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16832140 Supplier ProSci Llc Supplier No. 295330100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

SF1 Polyclonal Antibody specifically detects SF1 in Human, Mouse, Dog samples. It is validated for ELISA, Western Blot

Specifications

Antigen SF1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution SF1 antibody can be used for detection of SF1 by ELISA at 1:62500. SF1 antibody can be used for detection of SF1 by western blot at 1.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_004621
Gene Alias SF1, BBP, MBBP, ZFM1, ZNF162, D11S636, ZCCHC25
Host Species Rabbit
Purification Method Antibody is purified by peptide affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7536
Target Species Human, Mouse, Dog
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store SF1 antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Other
Show More Show Less