missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETDB2 Antibody, ProSci
Description
SETDB2 Polyclonal Antibody specifically detects SETDB2 in Human samples. It is validated for ELISA, Western Blot
Specifications
Specifications
| Antigen | SETDB2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 ug/mL |
| Conjugate | Unconjugated |
| Dilution | Peptide ELISA: antibody detection limit dilution 1:32000.Western Blot:Approx 90kDa band observed in Human Heart lysates (calculated MW of 81.9kDa according to NP_114121.2). Recommended concentration: 0.1-0.3ug/ml. |
| Formulation | Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing. |
| Gene Accession No. | NP_114121.2,NP_001153780.1 |
| Gene Alias | SETDB2, SET domain, bifurcated 2, C13orf4, CLLD8, CLLL8, DKFZp586I0123, DKFZp761J1217, KMT1F, OTTHUMP00000018418, chronic lymphocytic leukemia deletion region 8, chronic lymphocytic leukemia deletion region gene 8 protein, histone-lysine N-methyltransferase SETDB2, lysine N-methyltransferase 1F |
| Host Species | Goat |
| Show More |