missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SESN2 Recombinant Rabbit Monoclonal Antibody (PSH05-78)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562886
Description
Positive Control: K-562 cell lysate, HEK-293 cell lysate, 293T cell lysate, HCT 116 cell lysate, LoVo cell lysate, HepG2 cell lysate, Jurkat cell lysate, U-87 MG cell lysate, NIH/3T3 cell lysate, RAW264.7 cell lysate, C6 cell lysate, PC-12 cell lysate.Tissue Specificity: Low tissue specificitySubcellular Location: Cytoplasm.Sequence Similarities: 86% Mouse/RatPredicted band size: 54 kDa.
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.Specifications
| SESN2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P58004, P58043 | |
| SESN2 | |
| Synthetic peptide within human SESN2 aa 231-280. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| PSH05-78 | |
| Unconjugated | |
| SESN2 | |
| Hi95; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; RGD1566319; Ses2; SESN2; SEST2; sestrin 2; sestrin-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 230784, 502988, 83667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |