Learn More
Labconco™ Service Fixture Kits
Kit includes copper tubing, brass valve, knob handle and outlet.
$489.90 - $1,299.34
Specifications
For Use With (Equipment) Protector XLE Laboratory Hood
View More Specs
Products 5
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
09039047
View Documents Promotion Details
Labconco™
9809300
Nitrogen (NIT) Service Fixture Kit
Each for $489.90
Only null left
09039048
View Documents Promotion Details
Labconco™
9809700
Oxygen (OXY) Service Fixture Kit
Each for $712.58
Only null left
09039039
View Documents Promotion Details
Labconco™
9808300
Cold Water (CW) Service Fixture Kit
Each for $489.90
Only null left
09039042
View Documents Promotion Details
Labconco™
9808700
Gas (GAS) Service Fixture Kit
Each for $559.88
Only null left
09039049
View Documents Promotion Details
Labconco™
9827900
Cold Water (CW) GN Fixture Kit
Each for $1,299.34
Only null left
Description
Description
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and tubing, color-coded fixture knob.
Specifications
Specifications
Protector XLE Laboratory Hood