missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Service Fixture Kits
Kit includes copper tubing, brass valve, knob handle and outlet.
Supplier: Labconco™ 9809700
Description
- Max. working pressure of 40 psi.
- Kits mount on the right or left side of any Protector Premier, XStream, XL, or PVC Hood.
- Each kit includes one remotely-controlled service fixture with valve and tubing, color-coded fixture knob.
Chemical Identifiers
| Protector XLE Laboratory Hood |
Specifications
| Protector XLE Laboratory Hood | |
| 1.8 Kg | |
| Oxygen (OXY) Service Fixture Kit |
| 4 lb. | |
| 0.25 in. dia. Copper Tubing, Brass Valve and Green Knob, Connector | |
| Oxygen service |