missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Serum Vials, Clear
Made from premium Type I Borosilicate Glass, known for its durability and chemical resistance, this material is perfect for high-performance parenteral drug packaging.
Supplier: Fisherbrand™ 337401
Description
- ISO 9001:2015 Certified:
- Adhering to the highest international quality standards, ensuring consistent product excellence.
- Complete Batch Traceability:
- Full production traceability from raw materials to final product, ensuring accountability and quality control.
- Rigorous Quality Checks:
- Quality control at every stage of production, from raw material inspection to final packaging, ensuring product integrity.
Specifications
| Clear | |
| Borosilicate Glass (Type 1) | |
| Non-sterile | |
| 10 mL | |
| 800 | |
| Tubular with blowback crimp neck finish | |
| Stoppers and Caps (Not Included) | |
| 800 | |
| Serum Vial | |
| No |
| 22.75 mm | |
| No | |
| Single-use | |
| No | |
| 100 | |
| Autoclavable | |
| 50 mm | |
| ISO 9001:2015 | |
| 20 mm |