missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Serum Vials, Amber
Made from premium Type I Borosilicate Glass, known for its durability and chemical resistance, this material is perfect for high-performance parenteral drug packaging.
$253.15 - $1,593.24
Specifications
| Color | Amber |
|---|---|
| Material | Borosilicate Glass (Type 1) |
| Pre-slit | No |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
|
02993450
|
Fisherbrand™
337376 |
50 mL |
Case of 130 Each for $268.15
|
|
|||||
|
02993452
|
Fisherbrand™
337378 |
100 mL |
Case of 88 Each for $253.15
|
|
|||||
|
02993456
|
Fisherbrand™
337399 |
5 mL |
Case of 1200 Each for $1,593.24
|
|
|||||
|
02993457
|
Fisherbrand™
337400 |
10 mL |
Case of 800 Each for $1,080.74
|
|
|||||
|
02993460
|
Fisherbrand™
337405 |
20 mL |
Case of 576 Each for $861.47
|
|
|||||
Description
- ISO 9001:2015 Certified:
- Adhering to the highest international quality standards, ensuring consistent product excellence.
- Complete Batch Traceability:
- Full production traceability from raw materials to final product, ensuring accountability and quality control.
- Rigorous Quality Checks:
- Quality control at every stage of production, from raw material inspection to final packaging, ensuring product integrity.
Specifications
| Amber | |
| No | |
| Single-use | |
| Stoppers and Caps (Not Included) | |
| No | |
| ISO 9001:2015 |
| Borosilicate Glass (Type 1) | |
| Non-sterile | |
| No | |
| 20 mm | |
| Autoclavable | |
| Serum Vial |