missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertindole 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White |
| Quantity | 10 mg |
| Formula Weight | 440.95 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sertindole |
Safety and Handling
| RTECSNumber | NJ0621100 |
| TSCA | No |
| Recommended Storage | Refrigerator |