accessibility menu, dialog, popup

UserName

CAS RN 106516-24-9

FNNNNHOCl

CAS RN 106516-24-9

IUPAC Name: 1-(2-{4-[5-chloro-1-(4-fluorophenyl)-1H-indol-3-yl]piperidin-1-yl}ethyl)imidazolidin-2-one
Synonyms: sertindole sertindole (product) sertindolum sertindol sertindole hydrochloride SerLect C24H26ClFN4O serdolect sertindole (substance) sertindol [inn-spanish]
Molecular Weight (g/mol): 440.95
Molecular Formula: C24H26ClFN4O
InChi Key: GZKLJWGUPQBVJQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1C=C(C2CCN(CCN3CCNC3=O)CC2)C2=CC(Cl)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results