missing translation for 'onlineSavingsMsg'
Learn More

Sertindole 95.0+%, TCI America™

Catalog No. S094250MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S094250MG 50 mg
S094210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S094250MG Supplier TCI America Supplier No. S094250MG
Only null left

Chemical Identifiers

CAS 106516-24-9
Molecular Formula C24H26ClFN4O
Molecular Weight (g/mol) 440.95
MDL Number MFCD00867749
InChI Key GZKLJWGUPQBVJQ-UHFFFAOYSA-N
Synonym 1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl]-1-piperidinyl]ethyl]-2-imidazolidinone
PubChem CID 60149
ChEBI CHEBI:9122
IUPAC Name 1-(2-{4-[5-chloro-1-(4-fluorophenyl)-1H-indol-3-yl]piperidin-1-yl}ethyl)imidazolidin-2-one
SMILES FC1=CC=C(C=C1)N1C=C(C2CCN(CCN3CCNC3=O)CC2)C2=CC(Cl)=CC=C12

Specifications

Melting Point 155°C
Color White
Quantity 50 mg
Formula Weight 440.95
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sertindole
RTECSNumber NJ0621100
TSCA No
Recommended Storage Refrigerator