missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINF2 Monoclonal Antibody (H8)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565984
Description
SERPINF2 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.Chemical Identifiers
| SERPINF2 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08697 | |
| 100525254, 5345 | |
| Recombinant Alpha 2-Antiplasmin (a2PI). | |
| Antibody |
| H8 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Pig | |
| SERPINF2 | |
| A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
| SERPINF2 | |
| Primary | |
| Protein A/G |
Specifications
| SERPINF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08697 | |
| SERPINF2 | |
| Recombinant Alpha 2-Antiplasmin (a2PI). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| H8 | |
| Unconjugated | |
| SERPINF2 | |
| A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100525254, 5345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |