missing translation for 'onlineSavingsMsg'
Learn More

SERPINA3 293T Cell Overexpression Lysate (Denatured), Abnova

Catalog No. 89000202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89000202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89000202 Supplier Abnova Corporation Supplier No. H00000012T01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell transient overexpression lysate for SERPINA3

The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting.

  • Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
  • Transfected cell line: 293T
  • Plasmid: pCMV-SERPINA3 full-length
  • Host: Human
  • Theoretical molecular weight: 47.7kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

For Use With (Application) Western Blot
Cell Line 293T
Description SERPINA3 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_001085.4
Molecular Weight (g/mol) 47.7
pH Range 6.8
Plasmid pCMV-SERPINA3 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-SERPINA3 antibody (H00000012-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less