missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serpin A1 ELISA Kit DFS Item

ELISA

$880.00 - $880.00

Specifications

Accession Number P01009
Assay Range 2.05 to 500 ng/mL
Assay Sensitivity 2 ng/mL
Calibration Range 2 ng/mL to 500 ng/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
EHSERPINA1 DFS Item
View Documents
Invitrogen™
EHSERPINA1
96 Tests
Pack for $880.00
Only null left
 
Description

Description

The Human Serpin A1 ELISA quantitates Hu Serpin A1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Serpin A1. Principle of the method The Human Serpin A1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.
Specifications

Specifications

P01009
2 ng/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
A1A,A1AT,AAT,PI,PI1,PRO2275,alpha1AT
<10
HRP
Immunology
2°C to 8°C
alpha-1 Antitrypsin
4 hrs 45 mins
2.05 to 500 ng/mL
2 ng/mL to 500 ng/mL
HRP
Proteases, Peptidases & Inhibitors
This ELISA pair antibody detects human Serpin A1. Other species not determined.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
5265
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
PRO0684, A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT
1 hrs 20 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'