Learn More
Serotonin hydrochloride, 98%
CAS: 153-98-0 | C10H13ClN2O | 212.677 g/mol
Supplier: Thermo Scientific Chemicals B2126303
Description
Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSerotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.
Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.
Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.
Specifications
| Serotonin hydrochloride | |
| 167°C to 172°C | |
| C10H13ClN2O | |
| 1 g | |
| 3570963 | |
| 14,8463 | |
| Soluble in water,alcohol,hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform. | |
| C1=CC2=C(C=C1O)C(=CN2)CCN.Cl | |
| 212.677 | |
| 212.68 |
| 153-98-0 | |
| 0.99 | |
| MFCD00012686 | |
| UN2811 | |
| Hygroscopic; Light sensitive | |
| serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
| MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
| 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride | |
| 160436 | |
| 98% |
Chemical Identifiers
| 153-98-0 | |
| 212.677 | |
| MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
| 160436 | |
| C1=CC2=C(C=C1O)C(=CN2)CCN.Cl |
| C10H13ClN2O | |
| MFCD00012686 | |
| serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
| 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 244-464-4
RTECSNumber : NM2571000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only