accessibility menu, dialog, popup

UserName

CAS RN 153-98-0

IUPAC Name: hydrogen 3-(2-aminoethyl)-1H-indol-5-ol chloride
Synonyms: hydrogen serotonin chloride
Molecular Weight (g/mol): 212.68
Molecular Formula: C10H13ClN2O
InChi Key: MDIGAZPGKJFIAH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CNC2=CC=C(O)C=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)