missing translation for 'onlineSavingsMsg'
Learn More

Serotonin hydrochloride, 98%

Catalog No. AAB2126309
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2126309 10 g
AAB2126303 1 g
AAB2126306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2126309 Supplier Thermo Scientific Chemicals Supplier No. B2126309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 153-98-0 | C10H13ClN2O | 212.677 g/mol

Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Serotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.

Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10H13ClN2O
Molecular Weight (g/mol) 212.677
MDL Number MFCD00012686
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
PubChem CID 160436
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Specifications

Melting Point 167°C to 172°C
Assay Percent Range 0.99
Quantity 10 g
UN Number UN2811
Beilstein 3570963
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,8463
Solubility Information Soluble in water,alcohol,hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.
Formula Weight 212.68
Percent Purity 98%
Chemical Name or Material Serotonin hydrochloride
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 244-464-4
RTECSNumber NM2571000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only