missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine, USP, 98.5-101.5%, Spectrum™ Chemical
S1023, 56-45-1, C3H7NO3
Supplier: Spectrum Chemical Mfg Cor S102325KGBL
Description
Spectrum™ Chemical Serine, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 56-45-1 | |
| 0.001 | |
| Fiber Drum | |
| 25 kg | |
| MTCFGRXMJLQNBG-UHFFFAOYNA-N | |
| 2-amino-3-hydroxypropanoic acid | |
| 98.5 to 101.5% |
| 1 | |
| 0.002 | |
| C3H7NO3 | |
| +14.0° to +15.6° | |
| NC(CO)C(O)=O | |
| 105.09 | |
| USP |
Chemical Identifiers
| 56-45-1 | |
| 105.09 | |
| 2-amino-3-hydroxypropanoic acid |
| C3H7NO3 | |
| MTCFGRXMJLQNBG-UHFFFAOYNA-N | |
| NC(CO)C(O)=O |