missing translation for 'onlineSavingsMsg'
Learn More

Serine, USP, 98.5-101.5%, Spectrum™ Chemical

S1023, 56-45-1, C3H7NO3

Supplier:  Spectrum Chemical Mfg Cor S1023100GM

 View more versions of this product

Catalog No. 18606182

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Serine, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

56-45-1
0.001
Poly Bottle
100 g
MTCFGRXMJLQNBG-UHFFFAOYNA-N
2-amino-3-hydroxypropanoic acid
98.5 to 101.5%
1
0.002
C3H7NO3
+14.0° to +15.6°
NC(CO)C(O)=O
105.09
USP

Chemical Identifiers

56-45-1
105.09
2-amino-3-hydroxypropanoic acid
C3H7NO3
MTCFGRXMJLQNBG-UHFFFAOYNA-N
NC(CO)C(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions