missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SERCA1 ATPase Recombinant Rabbit Monoclonal Antibody (23GB4340)

Catalog No. MA552693
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552693 20 μL
MA552692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552693 Supplier Invitrogen™ Supplier No. MA552693
Only null left

Rabbit Recombinant Monoclonal Antibody

SERCA1 ATPase Recombinant Monoclonal Antibody for ICC/IF, Western Blot

ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.

Specifications

Antigen SERCA1 ATPase
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4340
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP2A1
Gene Accession No. O14983
Gene Alias adult Ca2+ ATPase; ATP2A; ATP2A1; ATP2A3; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA; SERCA ATPase; Serca1; SERCA1a; SR Ca(2+)-ATPase 1
Gene Symbols ATP2A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP2A1/SERCA1 (1-54AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 487
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less