missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophobic

Excellent for coupling oligos or proteins directly to the beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

$1,023.31 - $4,153.00

Specifications

Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) DNA Sample Prep, Protein Purification
Diameter (Metric) 1 μm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09981123
View Documents Promotion Details
Cytiva
65152105050250
15 mL
Each for $1,023.31
Only null left
 
09981124
View Documents Promotion Details
Cytiva
65152105050350
100 mL
Each for $4,153.00
Only null left
 
Description

Description

  • Double shell magnetite beads
  • . High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
Specifications

Specifications

5% solids (50mg/mL)
2°C to 8°C
DNA Sample Prep, Protein Purification
1 μm
Videos
SDS
Documents

Documents