missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophilic

Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

$727.81 - $3,725.00

Specifications

Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
Diameter (Metric) 1 μm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09981115
View Documents Promotion Details
Cytiva
24152105050250
15 mL
Each for $727.81
Only null left
 
09981116
View Documents Promotion Details
Cytiva
24152105050350
100 mL
Each for $3,725.00
Only null left
 
Description

Description

  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Specifications

Specifications

5% solids (50mg/mL)
2°C to 8°C
1 μm
Videos
SDS
Documents

Documents