missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophilic
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Specifications
Specifications
| Concentration | 5% solids (50mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | DNA Sample Prep |
| Quantity | 100 mL |
| Diameter (Metric) | 1 μm |