missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequenase Version 2.0 DNA Polymerase

Catalog No. 70775Z1000U
Encompass
Change view
Click to view available options
Quantity:
1000 U
200 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70775Z1000U 1000 U
70775Y200UN 200 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70775Z1000U Supplier Applied Biosystems™ Supplier No. 70775Z1000UN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sequenase™ Version 2.0 DNA Polymerase is a genetically engineered form of T7 DNA polymerase. Unlike the wild-type enzyme it has virtually no 3'→5' exonuclease activity.

Specifications

Concentration 13 units/μL
Description Sequenase Version 2.0 DNA Polymerase, 1000 Units, 13 units/μL Concentration, Genetically-Engineered form of T7 DNA Polymerase, 7.5 pH, 37°C Optimum Temperature
For Use With (Application) Sequencing
Polymerase Genetically-Engineered form of T7 DNA Polymerase
Product Type Sequenase Version 2.0 DNA Polymerase
Quantity 1000 U
Purity 0.95

For research use only