missing translation for 'onlineSavingsMsg'
Learn More

STERIS Septihol™ WFI Sterile Alcohol Solution

A ready to use 70% (v/v) USP Isopropyl Alcohol and 30% USP WFI solution that kills a range of Gram-positive and Gram-negative bacteria. Double bagged and gamma irradiated for use in aseptic areas.

$594.29 - $673.88

Specifications

Product Type Alcohol Solution
Sterility Non-sterile
Composition 70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI)
Container Type Plastic Bottle
Filtered Filtered
View More Specs

Products 2
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
19415108
View Documents
STERIS
625077WR
22 oz.
Case of 12 Each for $594.29
Only null left
 
19415109
View Documents
STERIS
625096WR
32 oz.
Case of 12 Each for $673.88
Only null left
 
Description

Description

  • A mixture of 70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI)
  • Removes residue from hard surfaces even from the most critical surfaces
  • Free of stabilizers and denaturants for quick, and complete evaporation
  • Can be used straight out of the trigger spray bottle
  • Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
  • Filtered to 0.2 micron
  • Gamma irradiated to ensure sterility for easy introduction in critical areas
  • Product is filled and double bagged in a Grade B (Class 100) facility to minimize the potential of particles being introduced during use
  • Each lot is documented by Certificates of Analysis that contain Endotoxin, Irradiation and Sterility results
Specifications

Specifications

Alcohol Solution
70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI)
Filtered
Residue Removal on Hard Surfaces
Non-sterile
Plastic Bottle
RTU
12/Cs.
Videos
SDS
Documents

Documents