missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS Septihol™ WFI Sterile Alcohol Solution
A ready to use 70% (v/v) USP Isopropyl Alcohol and 30% USP WFI solution that kills a range of Gram-positive and Gram-negative bacteria. Double bagged and gamma irradiated for use in aseptic areas.
Supplier: STERIS 625096WR
Description
- A mixture of 70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI)
- Removes residue from hard surfaces even from the most critical surfaces
- Free of stabilizers and denaturants for quick, and complete evaporation
- Can be used straight out of the trigger spray bottle
- Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
- Filtered to 0.2 micron
- Gamma irradiated to ensure sterility for easy introduction in critical areas
- Product is filled and double bagged in a Grade B (Class 100) facility to minimize the potential of particles being introduced during use
- Each lot is documented by Certificates of Analysis that contain Endotoxin, Irradiation and Sterility results
Specifications
| Non-sterile | |
| Plastic Bottle | |
| RTU | |
| 12/Cs. |
| 70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI) | |
| Filtered | |
| Residue Removal on Hard Surfaces | |
| 32 oz. |