missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Septa for 0.2 mL MicroAmp™ tubes (for the 310 Genetic Analyzer)

Catalog No. 4305547
Encompass
Change view
Click to view available options
Quantity:
480 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4305547 480 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4305547 Supplier Applied Biosystems™ Supplier No. 4305547

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These are septa covers for 0.2mL sample tubes, for use with the 310 Genetic Analyzer.

Specifications

Content And Storage Each package contains 480 septa.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Line MicroAmp
Product Type Septa
Quantity 480 Each
Shipping Condition Room Temperature
Tube Type Disposable Tube, Conical Tube, Assay Tube

For research use only