missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPINEO (TM) P 600, Spectrum™ Chemical
S2019
$245.27 - $245.27
Specifications
| Color | Opaque, White, Yellow tint |
|---|---|
| pH | 5 to 7 |
| Physical Form | Liquid |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical SEPINEO (TM) P 600Specifications
| Opaque, White, Yellow tint | |
| Liquid | |
| Amber Glass Bottle |
| 5 to 7 | |
| Reagent |