missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPINEO (TM) P 600, Spectrum™ Chemical
Description
Spectrum™ Chemical SEPINEO (TM) P 600
Specifications
Specifications
| Color | Opaque, White, Yellow Tint |
| pH | 5 to 7 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Grade | Reagent |