missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q

Catalog No. 45002928
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002928 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002928 Supplier Cytiva Supplier No. 17051010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for preparative protein separations in both research and industrial applications. Cytiva Q Sepharose™ Fast Flow Chromatography Media is a strong anion exchanger based on the well established Sepharose Fast Flow ion exchange platform.

  • Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Matrix is composed of crosslinked 6% agarose beads
  • Part of Sepharose Fast Flow ion exchange platform
  • High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
  • Available in range of pre-packed process development tools

  • Ligand: quaternary amine
  • Ion exchanger type: strong anion exchanger
  • Ionic capacity: 0.18 to 0.25mmol Cl-/mL medium
  • Binding capacity: 120mg HSA/mL medium
  • Pressure/Flow specification: 400 to 700cm/hr., 100kPa, XK 50/30 column, bed height 15cm

Capture and purification of proteins.

Specifications

Quantity 25 mL
For Use With (Application) For Preparative Protein Separations in Both Research and Industrial AppliCatIons
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol
Storage Requirements 4°C to 30°C
Product Type Q Sepharose Fast Flow Media
Flow Rate 400 to 700 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 5.9 in.
Height (Metric) Bed 15 cm
Ionic Capacity 0.18 to 0.24 mM Cl-/mL
Ion Exchange Type Strong AnIon Exchanger
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose with Dextran surface extenders
pH Range 2 to 12
Storage Buffer 20% Ethanol
Ligand Type Quaternary Amine
Show More Show Less