missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephaorose™ Fast Flow Anion Exchange Media: DEAE

Catalog No. 45002929
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002929 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002929 Supplier Cytiva Supplier No. 17070901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for preparative protein separations in both research and industrial applications. Cytiva Life Sciences™ Sephaorose™ Fast Flow Anion Exchange Media: DEAE ensures low levels of non-specific binding.

  • Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Matrix is composed of crosslinked 6% agarose beads
  • Part of Sepharose Fast Flow ion exchange platform
  • High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
  • Available in range of pre-packed process development tools

  • Ligand: Diethylaminoethyl
  • Ion exchanger type: weak anion exchanger
  • Ionic capacity: 0.11 to 0.16mmol Cl-/mL medium
  • Binding capacity: 110mg HSA/mL medium
  • Pressure/Flow specification: 300 to 600cm/hr., 100kPa, XK 50/30 column, bed height 15cm

Capture and purification of proteins.

Specifications

Quantity 500 mL
For Use With (Application) For Preparative Protein Separations in Both Research and Industrial AppliCatIons
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol
Storage Requirements 4°C to 30°C
Product Type DEAE Sepharose Fast Flow Media
Flow Rate 400 to 700 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 5.9 in.
Height (Metric) Bed 15 cm
Ionic Capacity 0.11 to 0.16 mM Cl-/mL
Ion Exchange Type Strong AnIon Exchanger
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Diethylaminoethyl
Show More Show Less