Learn More
Sensory Organs and Nervous System Detail Set
Catalog No. S64949
Change view
Click to view available options
Type:
Nervous System Detail Set
Sensory Organs Detail Set
Quantity:
10
11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
S64949 Nervous System Detail Set 11
S64950 Sensory Organs Detail Set 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S64949 Supplier Customer Channels Group (Ccg) Supplier No. 70800/EMD
Take a unique look at the senses.
With these slide sets, your students can study the senses like never before. The Sensory Organs Detail Set contains 10 slides and the Nervous System Detail Set contains 11 slides.
Sensory Organs Set
  • Tongue, rabbit, c.s. with taste buds
  • Touch corpuscles in human skin, c.s.
  • Olfactory epithelium, dog, c.s.
  • External and internal ear, l.s., with eardrum and cochlea
  • Eye, retina, human, c.s.
  • Eye, optic nerve, human, c.s.
  • Eye, c.s., through cornea, iris, and ciliary body
  • Eye, cornea, cow, c.s.
  • Eyelid, cat, c.s., showing meibomian gland
  • Eye, entrance of optic nerve in retina, c.s.
Nervous System Set
  • Cerebrum, human cortex, c.s.
  • Cerebellum, human, c.s. Weigert stained
  • Spinal cord, human, c.s.
  • Nerve, human, l.s.
  • Nerve, human, c.s.
  • Spinal cord, cat, c.s. Klazver-Barrera stained
  • Spinal cord, cow, c.s. Nissl stained
  • Cerebrum, cat, c.s. Golgi stained
  • Brain, rat, median l.s.
  • Vertebra with spinal cord, rat, c.s.
  • Cerebellum, human, c.s.
Specifications
Type Nervous System Detail Set
Quantity 11
Slide Type Prepared Slide