Learn More
Sensidyne™ Detector Tubes: Organic Gas Qualitative
Catalog No. 18073E
Change view
Click to view available options
Measurement Range:
NA
Catalog No. Measurement Range
18073E NA
1 options
Catalog No. 18073E Supplier Sensidyne™ Supplier No. 186B
Only null left
All detector tubes are sold in packs of 10.
Specifications
Components Measured Organic Gas Qualitative
Measurement Range NA
No. of Strokes 1+1
Quantity 10
Response Time 30+30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C