Learn More
Sensidyne™ Detector Tubes: Carbon Monoxide
Detector tubes
Supplier: Sensidyne™ 106S
Description
All detector tubes are sold in packs of 10.Chemical Identifiers
| Carbon Monoxide | |
| 10 |
Specifications
| Carbon Monoxide | |
| 3 | |
| 9 min. | |
| 0°C to 40°C |
| 10 to 250 | |
| 10 | |
| 32°F to 104°F |