Learn More
Sensidyne™ Detector Tubes: Ammonia
Catalog No. 18067B
Encompass
Change view
Click to view available options
Measurement Range:
0.2 to 20
0.5 to 30%
5 to 260
Response Time:
1 min.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
18067B 5 to 260 1 min.
18066Z 0.5 to 30% 1 min.
18067A 0.2 to 20 1 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18067B Supplier Sensidyne™ Supplier No. 105SC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.
  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10
Specifications
Components Measured Ammonia
Measurement Range 5 to 260
No. of Strokes 1
Quantity 10
Response Time 1 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C