missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Detector Tubes: Ammonia

Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.

Supplier:  Sensidyne™ 105SD

 View more versions of this product

Catalog No. 18067A


Only null left

Description

Description

  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10
Specifications

Chemical Identifiers

Ammonia
10
0°C to 40°C
1
32°F to 104°F

Specifications

Ammonia
1
1 min.
0°C to 40°C
0.2 to 20
10
32°F to 104°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions