missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Detector Tubes: Ammonia

Catalog No. 18066Z
Change view
Click to view available options
Measurement Range:
0.2 to 20
0.5 to 30%
5 to 260
Response Time:
1 min.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
18066Z 0.5 to 30% 1 min.
18067A 0.2 to 20 1 min.
18067B 5 to 260 1 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18066Z Supplier Sensidyne™ Supplier No. 105SH

Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.

  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10

Specifications

Components Measured Ammonia
Measurement Range 0.5 to 30%
No. of Strokes 1
Quantity 10
Response Time 1 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C