Learn More
Sensidyne™ Detector Tubes: Carbon Dioxide
Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.
Supplier:  Sensidyne™ 126SF
 View more versions of this product
Catalog No. 18067X
Only null left
Description
Description
  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10
Specifications
Chemical Identifiers
Carbon Dioxide
10
Specifications
Carbon Dioxide
n/a
2 min.
0°C to 40°C
100 to 4000
10
32°F to 104°F
Product Suggestions
Product Suggestions