missing translation for 'onlineSavingsMsg'
Learn More

Sensidyne™ Detector Tubes: Carbon Dioxide

Catalog No. 18067W
Change view
Click to view available options
Measurement Range:
0.1 to 5.2%
100 to 4000
5 to 50%
Response Time:
1.5 min.
2 min.
5 min.
No. of Strokes:
1/2
n/a
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time No. of Strokes
18067W 5 to 50% 1.5 min. 1/2
18067X 100 to 4000 2 min. n/a
18067Z 0.1 to 5.2% 5 min. n/a
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18067W Supplier Sensidyne™ Supplier No. 126UH

Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.

  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10

Specifications

Components Measured Carbon Dioxide
Measurement Range 5 to 50%
No. of Strokes 1/2
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C