missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sennoside D, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 37271-17-3 |
| Physical Form | Neat |
| Linear Formula | C42H40O19 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 848.76 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |