missing translation for 'onlineSavingsMsg'
Learn More

Sennoside A, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015606
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015606 5 mg
111015607 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015606 Supplier Merck Emd Millipore Supplier No. 689095MGF
Only null left

Sennoside A is a hydroxy anthracene glycoside generally present in the polyherbal formulations that contain Cassia angustifolia leaves, which is used in the treatment of constipation.

Featured Industries: Food and Beverages

Specifications

CAS 81-27-6
Physical Form Neat
Linear Formula C42H38O20
MDL Number MFCD00151527
Type Analytical Standard
Quantity 5 mg
Formula Weight 862.74
Percent Purity ≥96% (HPLC)
Grade Analytical Standard