missing translation for 'onlineSavingsMsg'
Learn More

Semicarbazide Hydrochloride 99.0+%, TCI America™

Catalog No. S003225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S003225G 25 g
S0032500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S003225G Supplier TCI America Supplier No. S003225G

Chemical Identifiers

CAS 563-41-7
Molecular Formula CH6ClN3O
Molecular Weight (g/mol) 111.53
MDL Number MFCD00013009
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
PubChem CID 11236
ChEBI CHEBI:82532
IUPAC Name hydrogen aminourea chloride
SMILES [H+].[Cl-].NNC(N)=O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 111.53
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Semicarbazide Hydrochloride
RTECSNumber VT3500000
TSCA Yes